missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetamido-3-nitrobenzoic acid, 97+%

$63.69 - $621.47

Chemical Identifiers

CAS 1539-06-6
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
MDL Number MFCD00014703
InChI Key BRQIMWBIZLRLSV-UHFFFAOYSA-N
Synonym 4-acetamido-3-nitrobenzoic acid, 4-acetylamino-3-nitrobenzoic acid, benzoic acid, 4-acetylamino-3-nitro, unii-9w2250fzkc, 4-acetamino-3-nitrobenzoic acid, 4-acetamido-3-nitro-benzoic acid, 4-acetylamino-3-nitrobenzoic acid, 4-acetylamino-3-nitro-benzoic acid, pubchem16827, acmc-1bqk5
PubChem CID 73758
SMILES CC(=O)NC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1036022 View Documents Alfa Aesar™
A1036022
100g Each for $621.47
 
AAA1036006 View Documents Alfa Aesar™
A1036006
5g Each for $63.69
 
AAA1036014 View Documents Alfa Aesar™
A1036014
25g Each for $208.19
 
Specifications

Chemical Identifiers

1539-06-6
224.172
BRQIMWBIZLRLSV-UHFFFAOYSA-N
73758
C9H8N2O5
MFCD00014703
4-acetamido-3-nitrobenzoic acid, 4-acetylamino-3-nitrobenzoic acid, benzoic acid, 4-acetylamino-3-nitro, unii-9w2250fzkc, 4-acetamino-3-nitrobenzoic acid, 4-acetamido-3-nitro-benzoic acid, 4-acetylamino-3-nitrobenzoic acid, 4-acetylamino-3-nitro-benzoic acid, pubchem16827, acmc-1bqk5
CC(=O)NC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

223°C to 228°C
1539-06-6
MFCD00014703
4-acetamido-3-nitrobenzoic acid, 4-acetylamino-3-nitrobenzoic acid, benzoic acid, 4-acetylamino-3-nitro, unii-9w2250fzkc, 4-acetamino-3-nitrobenzoic acid, 4-acetamido-3-nitro-benzoic acid, 4-acetylamino-3-nitrobenzoic acid, 4-acetylamino-3-nitro-benzoic acid, pubchem16827, acmc-1bqk5
CC(=O)NC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
73758
≥97%
100g
C9H8N2O5
2217819
BRQIMWBIZLRLSV-UHFFFAOYSA-N
224.172
224.17
4-Acetamido-3-nitrobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA