Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Acetamido-3'-bromoacetophenone, 98%

$102.61 - $384.23

Chemical Identifiers

Chemical Name or Material 4'-Acetamido-3'-bromoacetophenone
CAS 101209-08-9
Molecular Formula C10H10BrNO2
Formula Weight 256.1
MDL Number MFCD00051781
Beilstein 3272108
Synonym 4'-Acetyl-2'-bromoacetanilide; 2-Bromo-N,4-diacetylaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0762606

  SDS
alfa aesar™
L0762606
5g Each for $102.61

AAL0762614

  SDS
alfa aesar™
L0762614
25g Each for $384.23
Description & Specifications

Specifications

Formula Weight 256.1
CAS 101209-08-9
Chemical Name or Material 4'-Acetamido-3'-bromoacetophenone
Assay 98%
Molecular Formula C10H10BrNO2
MDL Number MFCD00051781
Synonym 4'-Acetyl-2'-bromoacetanilide; 2-Bromo-N,4-diacetylaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 3272108
Melting Point 139° to 141°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.