missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetamido-2-methylbenzonitrile, 97%

$96.30 - $309.41

Chemical Identifiers

CAS 321162-59-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.20
MDL Number MFCD02258877
InChI Key SRCREJLHAPEANJ-UHFFFAOYSA-N
Synonym 2-methyl-4-acetylamino benzonitrile, 4'-cyano-3'-methylacetanilide, 4-acetamido-2-methyl benzonitrile, 4-acetamido-2-methylbenzonitrile, acetamide,n-4-cyano-3-methylphenyl, acmc-20amqe, n-4-cyano-3-methylphenyl acetamide
PubChem CID 2737380
IUPAC Name N-(4-cyano-3-methylphenyl)acetamide
SMILES CC(=O)NC1=CC(C)=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2768606
View Documents
Alfa Aesar™
H2768606
5g
Each for $96.30
 
AAH2768614
View Documents
Alfa Aesar™
H2768614
25g
Each for $309.41
 
Description

Description

Specifications

Chemical Identifiers

321162-59-8
174.20
SRCREJLHAPEANJ-UHFFFAOYSA-N
2737380
CC(=O)NC1=CC(C)=C(C=C1)C#N
C10H10N2O
MFCD02258877
2-methyl-4-acetylamino benzonitrile, 4'-cyano-3'-methylacetanilide, 4-acetamido-2-methyl benzonitrile, 4-acetamido-2-methylbenzonitrile, acetamide,n-4-cyano-3-methylphenyl, acmc-20amqe, n-4-cyano-3-methylphenyl acetamide
N-(4-cyano-3-methylphenyl)acetamide

Specifications

161°C to 164°C
321162-59-8
MFCD02258877
2-methyl-4-acetylamino benzonitrile, 4'-cyano-3'-methylacetanilide, 4-acetamido-2-methyl benzonitrile, 4-acetamido-2-methylbenzonitrile, acetamide,n-4-cyano-3-methylphenyl, acmc-20amqe, n-4-cyano-3-methylphenyl acetamide
CC(=O)NC1=CC(C)=C(C=C1)C#N
174.20
174.2
4-Acetamido-2-methylbenzonitrile
5g
C10H10N2O
UN3439
SRCREJLHAPEANJ-UHFFFAOYSA-N
N-(4-cyano-3-methylphenyl)acetamide
2737380
97%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications