Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,9-Dioxa-1,12-dodecanediamine, 99%

Chemical Identifiers

Chemical Name or Material 4,9-Dioxa-1,12-dodecanediamine
CAS 7300-34-7
Molecular Formula C10H24N2O2
Linear Formula H2N(CH2)3O(CH2)4O(CH2)3NH2
Formula Weight 204.31
MDL Number MFCD00009802
UN Number UN2922
Beilstein 1750788
Synonym 1,4-Bis(3-aminopropoxy)butane; 1,12-Diamino-4,9-dioxadodecane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0956814

  SDS
alfa aesar™
L0956814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.31
Chemical Name or Material 4,9-Dioxa-1,12-dodecanediamine
CAS 7300-34-7
Molecular Formula C10H24N2O2
MDL Number MFCD00009802
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
H311-H314-H318-H332
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Odor Amine-like
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Melting Point 4° to 6°C
UN Number UN2922
Linear Formula H2N(CH2)3O(CH2)4O(CH2)3NH2
TSCA TSCA
Synonym 1,4-Bis(3-aminopropoxy)butane; 1,12-Diamino-4,9-dioxadodecane
Assay 99%
RTECS Number TX1515087
Health Hazard 1 Danger
Flash Point 159°C (318°F)
Quantity 25g
Boiling Point 298°C
Beilstein 1750788
pH 10
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H314-H318-H332
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.