Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(9-Carbazolyl)benzeneboronic acid pinacol ester, 95%

$72.32 - $296.84

Chemical Identifiers

Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid pinacol ester
Molecular Formula C24H24BNO2
Formula Weight 369.27
MDL Number MFCD16294549
Synonym 9-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5194803

  SDS
alfa aesar™
H5194803
1g Each for $72.32

AAH5194806

  SDS
alfa aesar™
H5194806
5g Each for $296.84
Description & Specifications

Specifications

Formula Weight 369.27
Quantity 1g
Molecular Formula C24H24BNO2
Synonym 9-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbazole
MDL Number MFCD16294549
Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid pinacol ester
Assay 95%

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.