Please login to your online account to display your discounted pricing

4μ8C, Tocris™

Inhibits IRE1α endoribonuclease activity

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material 4μ8C
Assay Percent Range >98%
Molecular Formula C11H8O4
Formula Weight 204.18
Synonym 7-Hydroxy-4-methyl-2-oxo-2H-1-benzopyran-8-carboxaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

447910

 
r & d systems
RDS447910
10mg Each for $221.01

447950

 
r & d systems
RDS447950
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 204.18
Quantity 10mg
Chemical Name or Material 4μ8C
Assay Percent Range >98%
Molecular Formula C11H8O4
CAS 14003-96-4
Synonym 7-Hydroxy-4-methyl-2-oxo-2H-1-benzopyran-8-carboxaldehyde
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C