missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,7-Diphenyl-1,10-phenanthroline, 99%, ACROS Organics™

Manufacturer:  Acros Organics 158320010

 View more versions of this product

Catalog No. AC158320010



Specifications

Specifications

4, 7-Diphenyl-1, 10-phenanthroline
Glass bottle
0.1% max.
218.0°C to 221.0°C
1662-01-7
100.0
C24H16N2
bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
DHDHJYNTEFLIHY-UHFFFAOYSA-N
4,7-diphenyl-1,10-phenanthroline
72812
332.39
99%
Authentic
For determination of iron: Passes Test
Pink to White
1g
98.5
99%
MFCD00004976
Solubility in water: insoluble.
C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5
332.406
CHEBI:77995
Crystalline Powder, Crystals or Crystalline Mass

Chemical Identifiers

1662-01-7
332.406
DHDHJYNTEFLIHY-UHFFFAOYSA-N
72812
4,7-diphenyl-1,10-phenanthroline
C24H16N2
MFCD00004976
bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
CHEBI:77995
C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5
SDS Documents