Please login to your online account to display your discounted pricing

4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid
CAS 175137-58-3
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H8N4O2
Formula Weight 192.18
MDL Number MFCD00067873
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM01115EA

  SDS
fisher scientific company
KM01115EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Assay 95%
Formula Weight 192.18
CAS Min % 95.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C8H8N4O2
MDL Number MFCD00067873
CAS 175137-58-3
Packaging Amber Glass Bottle
Chemical Name or Material 4,7-Dimethylpyrazolo[5,1-c][1,2,4]triazine-3-carboxylic acid
Assay Percent Range 95%
Health Hazard 1 Warning