Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,7-Dimethyl-5-decyn-4,7-diol, (+/-) + meso, 98+%

$25.76 - $103.66

Chemical Identifiers

Chemical Name or Material 4,7-Dimethyl-5-decyn-4,7-diol
CAS 126-87-4
Name Note (+/-) + meso
Molecular Formula C12H22O2
Formula Weight 198.31
MDL Number MFCD00021836
Beilstein 1723151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2055906

  SDS
alfa aesar™
B2055906
5g Each for $25.76

AAB2055914

  SDS
alfa aesar™
B2055914
25g Each for $103.66
Description & Specifications

Specifications

Formula Weight 198.31
CAS 126-87-4
Quantity 5g
Beilstein 1723151
Name Note (+/-) + meso
MDL Number MFCD00021836
Assay ≥98%
Melting Point 67° to 70°C
Molecular Formula C12H22O2
Chemical Name or Material 4,7-Dimethyl-5-decyn-4,7-diol
Boiling Point 125° to 127°C (3mmHg)