missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4',7-Dimethoxyisoflavone, 97%

Manufacturer:  Alfa Aesar™ L13838ME

 View more versions of this product

Catalog No. AAL13838ME



Specifications

Specifications

4',7-Dimethoxyisoflavone
500mg
C17H14O4
265550
LPNBCGIVZXHHHO-UHFFFAOYSA-N
7-methoxy-3-(4-methoxyphenyl)chromen-4-one
136419
97%
161°C to 163°C
1157-39-7
MFCD00075889
4',7-dimethoxyisoflavone, 7,4'-dimethoxyisoflavone, 7-methoxy-3-4-methoxyphenyl-4h-chromen-4-one, 7,4'-di-o-methyldaidzein, unii-f5547dw3co, 7-methoxy-3-4-methoxyphenyl chromen-4-one, daidzein dimethyl ether, spectrum_000697, specplus_000113, spectrum2_000185
COC1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)OC
282.295
282.3

Chemical Identifiers

1157-39-7
282.295
LPNBCGIVZXHHHO-UHFFFAOYSA-N
136419
COC1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)OC
C17H14O4
MFCD00075889
4',7-dimethoxyisoflavone, 7,4'-dimethoxyisoflavone, 7-methoxy-3-4-methoxyphenyl-4h-chromen-4-one, 7,4'-di-o-methyldaidzein, unii-f5547dw3co, 7-methoxy-3-4-methoxyphenyl chromen-4-one, daidzein dimethyl ether, spectrum_000697, specplus_000113, spectrum2_000185
7-methoxy-3-(4-methoxyphenyl)chromen-4-one
Documents