missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4',7-Dihydroxyisoflavone, 98+%, Thermo Scientific Chemicals

Analog of genistein and an agonist at estrogen recpetors

Supplier:  Thermo Scientific Chemicals J6376303

 View more versions of this product

Catalog No. AAJ6376303



Description

Description

Analog of genistein and an agonist at estrogen recpetors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4',7-Dihydroxyisoflavone
328°C to 332°C (decomposition)
C15H10O4
1 g
14,2801
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
5281708
254.24
Powder
486-66-8
White to Yellow
MFCD00016954
231523
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
254.241
CHEBI:28197
≥98%

Chemical Identifiers

486-66-8
254.241
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
5281708
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
C15H10O4
MFCD00016954
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
CHEBI:28197
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 207-635-4

RTECSNumber : DJ3100040

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only