missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,7-Dihydroxy-1,10-phenanthroline

Manufacturer:  Alfa Aesar™ 3143409

 View more versions of this product

Catalog No. AA3143409



Specifications

Specifications

4,7-Dihydroxy-1,10-phenanthroline
>300°C (decomposition)
3922-40-5
C12H8N2O2
1,10-Phenanthroline-4,7-diol
C1=CC2=C(C3=C1C(=O)C=CN3)NC=CC2=O
212.208
212.21
Yellow
10g
Hygroscopic
MFCD00004975
SLIBCJURSADKPV-UHFFFAOYSA-N
1,10-dihydro-1,10-phenanthroline-4,7-dione
77524
Powder

Chemical Identifiers

3922-40-5
212.208
SLIBCJURSADKPV-UHFFFAOYSA-N
77524
C1=CC2=C(C3=C1C(=O)C=CN3)NC=CC2=O
C12H8N2O2
MFCD00004975
1,10-Phenanthroline-4,7-diol
1,10-dihydro-1,10-phenanthroline-4,7-dione
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

SDS Documents