Alfa Aesar™ 4,7-Dichloroisatin, 98%
Manufacturer:  Alfa Aesar™ A1485314
 View more versions of this product
Catalog No. AAA1485314
Specifications
Specifications
4,7-Dichloroisatin
18711-13-2
C8H3Cl2NO2
168731
NUXYYWOWNFEMNH-UHFFFAOYSA-N
4,7-dichloro-1H-indole-2,3-dione
2734793
98%
246°C to 249°C
25g
MFCD00047214
4,7-dichloroisatin, 4,7-dichloroindoline-2,3-dione, 4,7-dichloro isatin, 4,7-dichloro-isatin, 4,7-dichloro-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 4,7-dichloro, 4,7-dichloroisatin;, 4,7-dichloroisatine;, d0ek7h, isatin-based compound, 51
C1=CC(=C2C(=C1Cl)C(=O)C(=O)N2)Cl
216.017
216.02
Chemical Identifiers
18711-13-2
216.017
NUXYYWOWNFEMNH-UHFFFAOYSA-N
2734793
C1=CC(=C2C(=C1Cl)C(=O)C(=O)N2)Cl
C8H3Cl2NO2
MFCD00047214
4,7-dichloroisatin, 4,7-dichloroindoline-2,3-dione, 4,7-dichloro isatin, 4,7-dichloro-isatin, 4,7-dichloro-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 4,7-dichloro, 4,7-dichloroisatin;, 4,7-dichloroisatine;, d0ek7h, isatin-based compound, 51
4,7-dichloro-1H-indole-2,3-dione
Documents