Learn More
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04450402
Description
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane is used as a phase transfer catalysts by transferring ions from one phase to another. It is involved in the synthesis of alkalides and electrides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane | |
Yellow | |
C16H32N2O5 | |
MFCD00005108 | |
616444 | |
4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane, cryptating agent 221, kryptofix 221, 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5. tricosane, cryptofix 221, kryptofix-221, kryptofix r 221, kryptofix™ 221 | |
HDLXPNDSLDLJHF-UHFFFAOYSA-N | |
4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane | |
123438 | |
97% |
31364-42-8 | |
1.11 g/mL | |
1.501 | |
0.5 g | |
Light sensitive | |
Miscible with water. | |
C1COCCN2CCOCCOCCN(CCO1)CCOCC2 | |
332.44 | |
332.44 | |
Liquid |
Chemical Identifiers
31364-42-8 | |
332.44 | |
HDLXPNDSLDLJHF-UHFFFAOYSA-N | |
123438 | |
C1COCCN2CCOCCOCCN(CCO1)CCOCC2 |
C16H32N2O5 | |
MFCD00005108 | |
4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane, cryptating agent 221, kryptofix 221, 4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5. tricosane, cryptofix 221, kryptofix-221, kryptofix r 221, kryptofix™ 221 | |
4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 250-592-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only