missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4450477

 View more versions of this product

Catalog No. AA4450477



Specifications

Specifications

4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
1.501
Yellow
31364-42-8
MFCD00005108
4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane
HDLXPNDSLDLJHF-UHFFFAOYSA-N
4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
123438
Liquid
1.11 g/mL
Light sensitive
0.1g
C16H32N2O5
616444
Miscible with water.
C1COCCN2CCOCCOCCN1CCOCCOCC2
332.441
332.44
97%

Chemical Identifiers

31364-42-8
332.441
HDLXPNDSLDLJHF-UHFFFAOYSA-N
123438
C1COCCN2CCOCCOCCN1CCOCCOCC2
C16H32N2O5
MFCD00005108
4,7,13,16,21-pentaoxa-1,10-diazabicyclo 8.8.5 tricosane
4,7,13,16,21-pentaoxa-1,10-diazabicyclo[8.8.5]tricosane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 250-592-1

TSCA : No

Recommended Storage : Keep cold

Documents