Please login to your online account to display your discounted pricing

4-[6-(Tributylstannyl)-2-pyridinyl]morpholine, 95%, Maybridge

$198.37 - $198.37

Chemical Identifiers

Chemical Name or Material 4-[6-(Tributylstannyl)-2-pyridinyl]morpholine
CAS 869901-24-6
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C21H38N2OSn
Formula Weight 453.25
MDL Number MFCD08271894
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC38341CB

 
fisher scientific uk ltd
CC38341CB
Amber Glass Bottle 250mg Each for $198.37

CC38341DA

 
fisher scientific company
CC38341DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 453.25
Quantity 250mg
Assay 95%
CAS Max % 100.0
Chemical Name or Material 4-[6-(Tributylstannyl)-2-pyridinyl]morpholine
Packaging Amber Glass Bottle
MDL Number MFCD08271894
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
CAS 869901-24-6
CAS Min % 94.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Molecular Formula C21H38N2OSn
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.