missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-O-Ethylidene-alpha-D-glucopyranose, TCI America™

Manufacturer:  TCI America E04021G

 View more versions of this product

Catalog No. E04021G



Specifications

Specifications

4,6-O-Ethylidene-alpha-D-glucopyranose
173°C
13224-99-2
MFCD00006820
VZPBLPQAMPVTFO-ACQOQSQZSA-N
(2R,4aR,6S,7R,8R,8aS)-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-6,7,8-triol
21581900
Crystal-Powder at 20°C
White-Yellow
1g
C8H14O6
4,6-o-ethylidene-a-d-glucose
CC1OCC2C(O1)C(C(C(O2)O)O)O
206.194
206.19
≥96.0% (GC)

Chemical Identifiers

13224-99-2
206.194
VZPBLPQAMPVTFO-ACQOQSQZSA-N
21581900
CC1OCC2C(O1)C(C(C(O2)O)O)O
C8H14O6
MFCD00006820
4,6-o-ethylidene-a-d-glucose
(2R,4aR,6S,7R,8R,8aS)-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-6,7,8-triol
Safety and Handling

Safety and Handling

TSCA : No

Documents