Please login to your online account to display your discounted pricing

4-(6-Isocyanatopyrid-2-yl)morpholine, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material 4-(6-Isocyanatopyrid-2-yl)morpholine
CAS 884507-15-7
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay Tech.
Molecular Formula C10H11N3O2
Formula Weight 205.22
MDL Number MFCD08435861
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC38306CB

  SDS
fisher scientific company
CC38306CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Quantity 250mg
Assay Tech.
Formula Weight 205.22
CAS Min % 80.0
CAS Max % 100.0
MDL Number MFCD08435861
CAS 884507-15-7
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-(6-Isocyanatopyrid-2-yl)morpholine
Molecular Formula C10H11N3O2
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Danger