missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dimethylpyrimidine, 94%

$61.69 - $153.58

Chemical Identifiers

CAS 1558-17-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006116
InChI Key LSBIUXKNVUBKRI-UHFFFAOYSA-N
Synonym 4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
PubChem CID 15257
IUPAC Name 4,6-dimethylpyrimidine
SMILES CC1=CC(=NC=N1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1030206 View Documents Alfa Aesar™
A1030206
5g Each for $153.58
 
AAA1030203 View Documents Alfa Aesar™
A1030203
1g Each for $61.69
 
Description

Description

Applications
4,6-Dimethylpyrimidine is used to produce (6-methyl-pyrimidin-4-yl)-pyruvic acid ethyl ester. It is used as pharmaceutical intermediate. It is also used in agrochemical and dyestuff field.

Solubility
Soluble in water at 20°C.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Chemical Identifiers

1558-17-4
108.144
LSBIUXKNVUBKRI-UHFFFAOYSA-N
15257
CC1=CC(=NC=N1)C
C6H8N2
MFCD00006116
4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
4,6-dimethylpyrimidine

Specifications

46°C (114°F)
153°C to 155°C
1558-17-4
MFCD00006116
107068
LSBIUXKNVUBKRI-UHFFFAOYSA-N
4,6-dimethylpyrimidine
15257
94%
Recognizable
5g
C6H8N2
UN1993
4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
CC1=CC(=NC=N1)C
108.144
108.14
4,6-Dimethylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA