Please login to your online account to display your discounted pricing

4,6-Dimethylpyrimidine, 93%, tech., ACROS Organics™

$147.91 - $147.91

Chemical Identifiers

Chemical Name or Material 4, 6-Dimethylpyrimidine
CAS 1558-17-4
CAS Min % 91.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥91%
Assay Percent Range 93%
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006116
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC116730025

  SDS
thermo fisher scientific
116730025
Glass bottle 2.5g Each for $147.91
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Liquid After Melting
Assay ≥91%
Formula Weight 108.14
Quantity 2.5g
Color Clear yellow
Beilstein 23, 95
Density 0.9800g/mL
CAS Min % 91.0
CAS Max % 100.0
CAS 1558-17-4
Chemical Name or Material 4, 6-Dimethylpyrimidine
Packaging Glass bottle
Refractive Index 1.4890 to 1.4930 (20°C, 589nm)
Assay Percent Range 93%
Molecular Formula C6H8N2
MDL Number MFCD00006116
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.