missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dimethylpyrimidine, 93%, tech., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 116730025

 View more versions of this product

Catalog No. AC116730025


 

Specifications Description & Specifications

Specifications

4, 6-Dimethylpyrimidine
Clear yellow
1558-17-4
100.0
Glass bottle
Technical Grade
≥91%
C6H8N2
MFCD00006116
0.9800g/mL
2.5g
91.0
Authentic
1.4890 to 1.4930 (20°C, 589nm)
Liquid After Melting
93%
108.14
23, 95

Chemical Identifiers

1558-17-4
100.0
C6H8N2
MFCD00006116
≥91%
91.0
93%
108.14
4, 6-Dimethylpyrimidine
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning