missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dimethyldibenzothiophene, 97%

Manufacturer:  Alfa Aesar™ H55472MD

 View more versions of this product

Catalog No. AAH55472MD



Description

Description

Applications
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

4,6-Dimethyldibenzothiophene
250mg
C14H12S
4,6-dimethyl dibenzo b thiophene
CC1=CC=CC2=C1SC3=C2C=CC=C3C
212.31
212.31
153°C to 157°C
1207-12-1
MFCD00216264
MYAQZIAVOLKEGW-UHFFFAOYSA-N
4,6-dimethyldibenzothiophene
1268103
97%

Chemical Identifiers

1207-12-1
212.31
MYAQZIAVOLKEGW-UHFFFAOYSA-N
1268103
CC1=CC=CC2=C1SC3=C2C=CC=C3C
C14H12S
MFCD00216264
4,6-dimethyl dibenzo b thiophene
4,6-dimethyldibenzothiophene
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 214-894-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents