missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,6-Dimethyldibenzothiophene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5547206
Description
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.
Notes
Incompatible with strong oxidizing agents.
Specifications
4,6-Dimethyldibenzothiophene | |
153°C to 157°C | |
MFCD00216264 | |
dibenzothiophene, 4,6-dimethyl, 4,6-dimethyldibenzo b,d thiophene, 4,6-dmdbt, 6,10-dimethyl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4,6-dimethyl-dibenzothiophen, ksc491i3n, bidd:gt0307, 4,6-dimethyl dibenzo b thiophene, 4,6-dimethyldibenzothiophene, 4,6-dimethylbenzo b benzo b thiophene | |
CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1 | |
212.31 | |
212.31 |
1207-12-1 | |
C14H12S | |
5 g | |
KMPJENUWHPZRGZ-UHFFFAOYSA-N | |
4,6-dimethyldibenzothiophene | |
1268103 | |
97% |
Chemical Identifiers
1207-12-1 | |
212.31 | |
KMPJENUWHPZRGZ-UHFFFAOYSA-N | |
1268103 | |
CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1 |
C14H12S | |
MFCD00216264 | |
dibenzothiophene, 4,6-dimethyl, 4,6-dimethyldibenzo b,d thiophene, 4,6-dmdbt, 6,10-dimethyl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4,6-dimethyl-dibenzothiophen, ksc491i3n, bidd:gt0307, 4,6-dimethyl dibenzo b thiophene, 4,6-dimethyldibenzothiophene, 4,6-dimethylbenzo b benzo b thiophene | |
4,6-dimethyldibenzothiophene |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 214-894-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only