missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,6-Dimethyl-2-pyrone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 295350010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4, 6-Dimethyl-2-pyrone | |
675-09-2 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00075555 | |
4,6-dimethyl-2-pyrone, 4,6-dimethyl-2h-pyran-2-one, mesitene lactone, 4,6-dimethylcoumalin, 2h-pyran-2-one, 4,6-dimethyl, 4,6-dimethyl-alpha-pyrone, 4,6-dimethyl-pyran-2-one, 2,4-dimethyl-alpha-pyrone, 2,4-dimethyl-.alpha.-pyrone, 4,6-dimethyl-.alpha.-pyrone | |
124.14 |
98% | |
97.5 | |
>110°C | |
97.5% min. (GC) | |
C7H8O2 | |
1g | |
124.14 | |
98% |
Chemical Identifiers
675-09-2 | |
124.14 | |
4,6-dimethyl-2-pyrone, 4,6-dimethyl-2h-pyran-2-one, mesitene lactone, 4,6-dimethylcoumalin, 2h-pyran-2-one, 4,6-dimethyl, 4,6-dimethyl-alpha-pyrone, 4,6-dimethyl-pyran-2-one, 2,4-dimethyl-alpha-pyrone, 2,4-dimethyl-.alpha.-pyrone, 4,6-dimethyl-.alpha.-pyrone |
C7H8O2 | |
MFCD00075555 |
Safety and Handling
EINECSNumber : 211-618-7
RTECSNumber : UQ0700000
TSCA : TSCA
Recommended Storage : Product may darken during storage
RUO â Research Use Only