missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dimethyl-2-nitroaniline, 97%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 442440050

Catalog No. AC442440050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4, 6-Dimethyl-2-nitroaniline
96% min. (HPLC)
5g
VSRYYONYIUUFFY-UHFFFAOYSA-N
2,4-dimethyl-6-nitroaniline
74219
97%
1635-84-3
C8H10N2O2
4,6-dimethyl-2-nitroaniline, 6-nitro-2,4-xylidine, 2,4-dimethyl-6-nitrophenyl amine, benzenamine, 2,4-dimethyl-6-nitro, 2,4-dimethyl-6-nitrophenylamine, acmc-209uyq, intermediates-zcf02230, 4-amino-5-nitro-m-xylene, cambridge id 5100320, 2-nitro-4,6-dimethylaniline
CC1=CC(=C(C(=C1)[N+](=O)[O-])N)C
166.18
166.18

Chemical Identifiers

1635-84-3
166.18
4,6-dimethyl-2-nitroaniline, 6-nitro-2,4-xylidine, 2,4-dimethyl-6-nitrophenyl amine, benzenamine, 2,4-dimethyl-6-nitro, 2,4-dimethyl-6-nitrophenylamine, acmc-209uyq, intermediates-zcf02230, 4-amino-5-nitro-m-xylene, cambridge id 5100320, 2-nitro-4,6-dimethylaniline
2,4-dimethyl-6-nitroaniline
C8H10N2O2
VSRYYONYIUUFFY-UHFFFAOYSA-N
74219
CC1=CC(=C(C(=C1)[N+](=O)[O-])N)C
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

EINECSNumber : 216-662-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only