missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dimethyl-2-(methylsulfonyl)pyrimidine, 98%

Manufacturer:  Alfa Aesar™ H6402922

 View more versions of this product

Catalog No. AAH6402922



Specifications

Specifications

4,6-Dimethyl-2-(methylsulfonyl)pyrimidine
80°C to 85°C (literature)
35144-22-0
MFCD03788286
4,6-dimethyl-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethylpyrimidine, 2-methanesulfonyl-4,6-dimethyl-pyrimidine, 2-methylsulfonyl-4,6-dimethylpyrimidine, pyrimidine, 4,6-dimethyl-2-methylsulfonyl, 4,6-dimethyl-2-methylsulfonyl-pyrimidine, pubchem10458, pubchem21462, acmc-209idk, ksc496i3d
CC1=CC(=NC(=N1)S(=O)(=O)C)C
186.229
186.23
98%
Red to Brown
100g
C7H10N2O2S
778406
ZHPSNGCLCHWTRG-UHFFFAOYSA-N
4,6-dimethyl-2-methylsulfonylpyrimidine
2772382
Crystalline Solid

Chemical Identifiers

35144-22-0
186.229
ZHPSNGCLCHWTRG-UHFFFAOYSA-N
2772382
CC1=CC(=NC(=N1)S(=O)(=O)C)C
C7H10N2O2S
MFCD03788286
4,6-dimethyl-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethylpyrimidine, 2-methanesulfonyl-4,6-dimethyl-pyrimidine, 2-methylsulfonyl-4,6-dimethylpyrimidine, pyrimidine, 4,6-dimethyl-2-methylsulfonyl, 4,6-dimethyl-2-methylsulfonyl-pyrimidine, pubchem10458, pubchem21462, acmc-209idk, ksc496i3d
4,6-dimethyl-2-methylsulfonylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents