Please login to your online account to display your discounted pricing

4,6-Dimethyl-1,3,5-triazin-2-amine hydrate, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate
CAS 175278-59-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C5H8N4·H2O
Formula Weight 142.17
MDL Number MFCD00052756
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF02292EA

 
fisher scientific company
RF02292EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.17
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate
MDL Number MFCD00052756
CAS 175278-59-8
Assay Percent Range 95%+
Molecular Formula C5H8N4·H2O
Packaging Amber Glass Bottle