Please login to your online account to display your discounted pricing

4,6-Dimethyl-1,3,5-triazin-2-amine hydrate, ≥95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RF02292EA

 View more versions of this product

Catalog No. RF02292EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 142.17
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate
MDL Number MFCD00052756
CAS 175278-59-8
Assay Percent Range 95%+
Molecular Formula C5H8N4·H2O
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4,6-Dimethyl-1,3,5-triazin-2-amine hydrate
CAS 175278-59-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C5H8N4·H2O
Formula Weight 142.17
MDL Number MFCD00052756