Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine, 96%

$33.76 - $325.46

Chemical Identifiers

Chemical Name or Material 4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine
CAS 113583-35-0
Molecular Formula C7H10N2O4S
Formula Weight 218.23
MDL Number MFCD00672151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2700303

  SDS
alfa aesar™
H2700303
1g Each for $33.76

AAH2700306

  SDS
alfa aesar™
H2700306
5g Each for $91.61

AAH2700314

  SDS
alfa aesar™
H2700314
25g Each for $325.46
Description & Specifications

Specifications

Formula Weight 218.23
Chemical Name or Material 4,6-Dimethoxy-2-(methylsulfonyl)pyrimidine
Assay 96%
Molecular Formula C7H10N2O4S
MDL Number MFCD00672151
CAS 113583-35-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 129° to 133°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.