Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol, 97%

$292.46 - $292.46

Chemical Identifiers

Chemical Name or Material 4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol
CAS 93892-40-1
Molecular Formula C19H30O
Formula Weight 274.45
MDL Number MFCD00061045
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2089106

 
alfa aesar™
B2089106
5g Each for $292.46
Description & Specifications

Specifications

Formula Weight 274.45
Chemical Name or Material 4,6-Diisopropyl-1,1,3,3-tetramethyl-5-indanol
Melting Point 146° to 148°C
Assay 97%
Molecular Formula C19H30O
Quantity 5g
MDL Number MFCD00061045
CAS 93892-40-1