missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dihydroxypyrimidine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC115085000

 View more versions of this product

Catalog No. AC115085000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4, 6-Dihydroxypyrimidine
Plastic bottle
>300°C
1193-24-4
100.0
C4H4N2O2
26,444
Solubility in water: 2.5g/L water (20° c)
OC1=CC(=O)NC=N1
112.09
112.09
98%
Authentic
Yellow to Orange
500g
97.5
98%
MFCD00016733
4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 4,6-dihydroxypyrimidin, 4,6-dihydroxypyrimidine, 4,6-pyrimidinediol, 6-hydroxy-1h-pyrimidin-4-one, 6-hydroxy-4 1h-pyrimidinone, 6-hydroxy-4-pyrimidinone, 6-hydroxypyrimidin-4 3h-one, pyrimidine-4,6-diol
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4-hydroxy-1H-pyrimidin-6-one
14512
Powder

Chemical Identifiers

1193-24-4
112.09
DUFGYCAXVIUXIP-UHFFFAOYSA-N
14512
OC1=CC(=O)NC=N1
C4H4N2O2
MFCD00016733
4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 4,6-dihydroxypyrimidin, 4,6-dihydroxypyrimidine, 4,6-pyrimidinediol, 6-hydroxy-1h-pyrimidin-4-one, 6-hydroxy-4 1h-pyrimidinone, 6-hydroxy-4-pyrimidinone, 6-hydroxypyrimidin-4 3h-one, pyrimidine-4,6-diol
4-hydroxy-1H-pyrimidin-6-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-772-3

SDS Documents