missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dihydroxypyrimidine, 98%

Manufacturer:  Alfa Aesar™ A1568822

 View more versions of this product

Catalog No. AAA1568822



Specifications

Specifications

4,6-Dihydroxypyrimidine
100g
C4H4N2O2
606433
DUFGYCAXVIUXIP-UHFFFAOYSA-N
4-hydroxy-1H-pyrimidin-6-one
14512
98%
>300°C
1193-24-4
MFCD00016733
4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
C1=C(N=CNC1=O)O
112.088
112.09

Chemical Identifiers

1193-24-4
112.088
DUFGYCAXVIUXIP-UHFFFAOYSA-N
14512
C1=C(N=CNC1=O)O
C4H4N2O2
MFCD00016733
4,6-dihydroxypyrimidine, pyrimidine-4,6-diol, 4,6-pyrimidinediol, 6-hydroxy-4 1h-pyrimidinone, 4 1h-pyrimidinone, 6-hydroxy, 4 3h-pyrimidinone, 6-hydroxy, 6-hydroxypyrimidin-4 3h-one, 6-hydroxy-1h-pyrimidin-4-one, 4,6-dihydroxypyrimidin, 6-hydroxy-4-pyrimidinone
4-hydroxy-1H-pyrimidin-6-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UW7523000

TSCA : TSCA

Documents