Learn More
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 339940050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine | |
98.5 | |
Authentic | |
Glass bottle | |
MFCD00056934 | |
oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van | |
C1=C2C(=NC(=O)NC2=O)NN1 | |
152.11 | |
CHEBI:28315 | |
99% |
2465-59-0 | |
100.0 | |
99% | |
C5H4N4O2 | |
5g | |
HXNFUBHNUDHIGC-UHFFFAOYSA-N | |
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione | |
4644 | |
152.11 |
Chemical Identifiers
2465-59-0 | |
152.11 | |
HXNFUBHNUDHIGC-UHFFFAOYSA-N | |
4644 | |
1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione |
C5H4N4O2 | |
MFCD00056934 | |
oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van | |
CHEBI:28315 | |
C1=C2C(=NC(=O)NC2=O)NN1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Danger
EINECSNumber : 219-570-9
RUO â Research Use Only