Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dihydroxy-5-nitropyrimidine, 98%

$31.41 - $103.14

Chemical Identifiers

Chemical Name or Material 4,6-Dihydroxy-5-nitropyrimidine
CAS 2164-83-2
Molecular Formula C4H3N3O4
Formula Weight 157.09
MDL Number MFCD00006110
Beilstein 611622
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2131206

 
alfa aesar™
B2131206
5g Each for $31.41

AAB2131214

 
alfa aesar™
B2131214
25g Each for $103.14
Description & Specifications

Specifications

Formula Weight 157.09
Chemical Name or Material 4,6-Dihydroxy-5-nitropyrimidine
TSCA TSCA
Assay 98%
Molecular Formula C4H3N3O4
MDL Number MFCD00006110
Beilstein 611622
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >300°C
Quantity 5g
CAS 2164-83-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.