Please login to your online account to display your discounted pricing

4,6-Dihydroxy-5-nitropyrimidine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4, 6-Dihydroxy-5-nitropyrimidine
Name Note 95%
CAS 2164-83-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C4H3N3O4
Formula Weight 157.09
MDL Number MFCD00006110
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305410050

  SDS
thermo fisher scientific
AC305410050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC305410250

  SDS
thermo fisher scientific
AC305410250
Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 157.09
Quantity 5g
Color Yellow
EINECS Number 218-504-6
TSCA TSCA
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym 5-Nitro-4, 6-pyrimidinediol
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4, 6-Dihydroxy-5-nitropyrimidine
MDL Number MFCD00006110
Molecular Formula C4H3N3O4
Assay Percent Range 95%
Name Note 95%
CAS 2164-83-2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.