Please login to your online account to display your discounted pricing

4,6-Dihydroxy-5-nitropyrimidine, 95%, ACROS Organics™


Quantity 25g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC305410250

 View more versions of this product

Catalog No. AC305410250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 157.09
Assay ≥94%
Physical Form Crystalline Powder
Quantity 25g
Color Yellow
TSCA TSCA
EINECS Number 218-504-6
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Synonym 5-Nitro-4, 6-pyrimidinediol
Chemical Name or Material 4, 6-Dihydroxy-5-nitropyrimidine
Molecular Formula C4H3N3O4
Name Note 95%
Assay Percent Range 95%
MDL Number MFCD00006110
CAS 2164-83-2

Specifications

Chemical Name or Material 4, 6-Dihydroxy-5-nitropyrimidine
Name Note 95%
CAS 2164-83-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C4H3N3O4
Formula Weight 157.09
MDL Number MFCD00006110
Synonym 5-Nitro-4, 6-pyrimidinediol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents