missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dihydroxy-2-methylpyrimidine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1053518

 View more versions of this product

Catalog No. AAA1053518



Description

Description

4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dihydroxy-2-methylpyrimidine is used in the preparation of 4,6-dichloro-2-methyl-pyrimidine.

Solubility
Soluble in sodium hydroxide.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

4,6-Dihydroxy-2-methylpyrimidine
>360°C
MFCD00006104,MFCD00205627,MFCD20486784
115646
Soluble in sodium hydroxide.
CC1=NC(O)=CC(=O)N1
126.12
126.12
40497-30-1
C5H6N2O2
50 g
4,6-dihydroxy-2-methylpyrimidine, 2-methylpyrimidine-4,6-diol, 2-methyl-4,6-dihydroxypyrimidine, 2-methyl-4,6-pyrimidinediol, 2-methyl-1h,5h-pyrimidine-4,5-dione, 4 1h-pyrimidinone, 6-hydroxy-2-methyl, 2-methyl-4,6-dihydroxy pyrimidine, 6-hydroxy-2-methyl-3h-pyrimidin-4-one, 6-hydroxy-2-methylpyrimidin-4 3h-one, 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one
BPSGVKFIQZZFNH-UHFFFAOYSA-N
4-hydroxy-2-methyl-1H-pyrimidin-6-one
222672
99%

Chemical Identifiers

40497-30-1
126.12
BPSGVKFIQZZFNH-UHFFFAOYSA-N
222672
CC1=NC(O)=CC(=O)N1
C5H6N2O2
MFCD00006104,MFCD00205627,MFCD20486784
4,6-dihydroxy-2-methylpyrimidine, 2-methylpyrimidine-4,6-diol, 2-methyl-4,6-dihydroxypyrimidine, 2-methyl-4,6-pyrimidinediol, 2-methyl-1h,5h-pyrimidine-4,5-dione, 4 1h-pyrimidinone, 6-hydroxy-2-methyl, 2-methyl-4,6-dihydroxy pyrimidine, 6-hydroxy-2-methyl-3h-pyrimidin-4-one, 6-hydroxy-2-methylpyrimidin-4 3h-one, 6-hydroxy-2-methyl-3,4-dihydropyrimidin-4-one
4-hydroxy-2-methyl-1H-pyrimidin-6-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 254-941-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only