missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dihydroxy-2-methylmercaptopyrimidine, 97%, ACROS Organics™

$64.47 - $64.47

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C5H6N2O2S
Formula Weight 158.17
Synonym 2-Methylthio-4, 6-pyrimidinediol
Chemical Name or Material 4, 6-Dihydroxy-2-methylmercaptopyrimidine
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC114970050 SDS Thermo Fisher Scientific
AC114970050
5g Each for $64.47
AC114970010 SDS Thermo Fisher Scientific
AC114970010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
158.17
4, 6-Dihydroxy-2-methylmercaptopyrimidine
C5H6N2O2S
2-Methylthio-4, 6-pyrimidinediol
≥96.0%

Specifications

5g
Authentic
97%
158.17
2-Methylthio-4, 6-pyrimidinediol
≥96.0%
1979-98-2
Powder
C5H6N2O2S
White
4, 6-Dihydroxy-2-methylmercaptopyrimidine

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.