Learn More
4,6-Difluoroindole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5453603
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Sparingly Soluble (0.062 g/L) (25°C), Calc.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
Specifications
4,6-Difluoroindole | |
C8H5F2N | |
1 g | |
4,6-difluoroindole, 1h-indole, 4,6-difluoro, 4,6-difluoro-indole, pubchem7179, acmc-209f4a, 1h-indole,4,6-difluoro, 1h-indole,4,6-difluoro-9ci, 4,6-difluoroindole 153.13, 4,6-difluoro-1h-indole; 1h-indole,4,6-difluoro-9ci | |
MHICCULQVCEWFH-UHFFFAOYSA-N | |
4,6-difluoro-1H-indole | |
2778736 | |
97% |
199526-97-1 | |
MFCD01075213 | |
Air sensitive | |
Sparingly Soluble (0.062g/L) (25°C),Calc. | |
C1=CNC2=CC(=CC(=C21)F)F | |
153.132 | |
153.13 |
Chemical Identifiers
199526-97-1 | |
153.132 | |
MHICCULQVCEWFH-UHFFFAOYSA-N | |
4,6-difluoro-1H-indole |
C8H5F2N | |
MFCD01075213 | |
2778736 | |
C1=CNC2=CC(=CC(=C21)F)F |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only