Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dichlorosalicylaldehyde, 98%

$136.31 - $136.31

Chemical Identifiers

Chemical Name or Material 4,6-Dichlorosalicylaldehyde
CAS 78443-72-8
Molecular Formula C7H4Cl2O2
Formula Weight 191.01g/mol
MDL Number MFCD00016590
Synonym 2,4-Dichloro-6-hydroxybenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2920406

 
alfa aesar™
H29204-06
5g Each for $136.31
Description & Specifications

Specifications

Melting Point 50° to 53°C
Quantity 5g
Chemical Name or Material 4,6-Dichlorosalicylaldehyde
CAS 78443-72-8
Assay 98%
Molecular Formula C7H4Cl2O2
Formula Weight 191.01g/mol
MDL Number MFCD00016590
Synonym 2,4-Dichloro-6-hydroxybenzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
H nmr spectral parameters are reported for salicylaldehyde and 15 of its derivatives in dilute CCl4 solutions, a positive spin-spin coupling is observed between the two sidechain protons.

Solubility
Sparingly Soluble (0.092 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.