Learn More
4,6-Dichloropyrimidine-5-carboxylic acid, 95%, Thermo Scientific Chemicals
$392.01 - $392.01
Chemical Identifiers
CAS | 87600-98-4 |
---|---|
Molecular Formula | C5H2Cl2N2O2 |
Molecular Weight (g/mol) | 192.983 |
MDL Number | MFCD09909611 |
InChI Key | KGBLYFGHJNGQBK-UHFFFAOYSA-N |
Synonym | 4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro |
PubChem CID | 24820487 |
IUPAC Name | 4,6-dichloropyrimidine-5-carboxylic acid |
SMILES | C1=NC(=C(C(=N1)Cl)C(=O)O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6397606
|
Thermo Scientific Chemicals
H6397606 |
5 g |
N/A
|
|
|||||
Description
4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
87600-98-4 | |
192.983 | |
KGBLYFGHJNGQBK-UHFFFAOYSA-N | |
24820487 | |
C1=NC(=C(C(=N1)Cl)C(=O)O)Cl |
C5H2Cl2N2O2 | |
MFCD09909611 | |
4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro | |
4,6-dichloropyrimidine-5-carboxylic acid |
Specifications
87600-98-4 | |
C5H2Cl2N2O2 | |
5 g | |
Slightly soluble in water. | |
C1=NC(=C(C(=N1)Cl)C(=O)O)Cl | |
192.983 | |
192.99 | |
Solid |
White | |
MFCD09909611 | |
4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro | |
KGBLYFGHJNGQBK-UHFFFAOYSA-N | |
4,6-dichloropyrimidine-5-carboxylic acid | |
24820487 | |
95% | |
4,6-Dichloropyrimidine-5-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only