missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dichloropyrimidine-5-carboxylic acid, 95%

Manufacturer:  Alfa Aesar™ H63976MD

 View more versions of this product

Catalog No. AAH63976MD



Description

Description

Applications
4,6-Dichloropyrimidine-5-carboxylic acid is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

4,6-Dichloropyrimidine-5-carboxylic acid
250mg
C5H2Cl2N2O2
4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylic acid
C1=NC(=C(C(=N1)Cl)C(=O)O)Cl
192.983
192.99
95%
White
87600-98-4
MFCD09909611
KGBLYFGHJNGQBK-UHFFFAOYSA-N
4,6-dichloropyrimidine-5-carboxylic acid
24820487
Solid

Chemical Identifiers

87600-98-4
192.983
KGBLYFGHJNGQBK-UHFFFAOYSA-N
24820487
C1=NC(=C(C(=N1)Cl)C(=O)O)Cl
C5H2Cl2N2O2
MFCD09909611
4,6-dichloro-5-pyrimidinecarboxylic acid, 4,6-dichloro-pyrimidine-5-carboxylic acid, 5-carboxy-4,6-dichloropyrimidine, 5-pyrimidinecarboxylic acid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylicacid, ksc446k2r, 4,6-dichloropyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 4,6-dichloro, 4,6-dichloro-pyrimidine-5-carboxylic acid
4,6-dichloropyrimidine-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents