missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dichloropyrimidine-5-carboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ H5444914

 View more versions of this product

Catalog No. AAH5444914



Description

Description

Applications
4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.

Notes
Incompatible with strong oxidizing agents and strong acids. Store in cool place.
Specifications

Specifications

4,6-Dichloropyrimidine-5-carboxaldehyde
25g
C5H2Cl2N2O
4,6-di chloro pyrimidine-5-aldehyde
C1=NC(=C(C(=N1)Cl)C=O)Cl
176.984
175.99
66°C to 71°C
5305-40-8
MFCD02257701
XQSJHQXYQAUDFC-UHFFFAOYSA-N
4,6-dichloropyrimidine-5-carbaldehyde
819691
97%

Chemical Identifiers

5305-40-8
176.984
XQSJHQXYQAUDFC-UHFFFAOYSA-N
819691
C1=NC(=C(C(=N1)Cl)C=O)Cl
C5H2Cl2N2O
MFCD02257701
4,6-di chloro pyrimidine-5-aldehyde
4,6-dichloropyrimidine-5-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

Documents