Learn More
4,6-Dichloropyrimidine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5444903
Description
4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,6-Dichloropyrimidine-5-carboxaldehyde is used as a substrate in the synthesis of N-terminal surrogate in amino acid and peptide analogues.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in cool place.
Specifications
4,6-Dichloropyrimidine-5-carboxaldehyde | |
66°C to 71°C | |
MFCD02257701 | |
4,6-dichloro-5-pyrimidinecarbaldehyde, 4,6-dichloro-pyrimidine-5-carbaldehyde, 4,6-dichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 4,6-dichloro, 4,6-dichloro-5-formylpyrimidine, 4,6-dichloro-5-pyrimidinecarboxaldehyde, 4,6-di chloro pyrimidine-5-aldehyde, 4,6-dichlorpyrimidin-5-carbaldehyd, zlchem 524, pubchem2117 | |
ClC1=NC=NC(Cl)=C1C=O | |
176.98 | |
175.99 |
5305-40-8 | |
C5H2Cl2N2O | |
1 g | |
XQSJHQXYQAUDFC-UHFFFAOYSA-N | |
4,6-dichloropyrimidine-5-carbaldehyde | |
819691 | |
97% |
Chemical Identifiers
5305-40-8 | |
176.98 | |
XQSJHQXYQAUDFC-UHFFFAOYSA-N | |
819691 | |
ClC1=NC=NC(Cl)=C1C=O |
C5H2Cl2N2O | |
MFCD02257701 | |
4,6-dichloro-5-pyrimidinecarbaldehyde, 4,6-dichloro-pyrimidine-5-carbaldehyde, 4,6-dichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 4,6-dichloro, 4,6-dichloro-5-formylpyrimidine, 4,6-dichloro-5-pyrimidinecarboxaldehyde, 4,6-di chloro pyrimidine-5-aldehyde, 4,6-dichlorpyrimidin-5-carbaldehyd, zlchem 524, pubchem2117 | |
4,6-dichloropyrimidine-5-carbaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only