Learn More
4,6-Dichloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6177003
Description
4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
4,6-Dichloro-5-fluoropyrimidine | |
50°C to 60°C (14 mmHg) | |
C4HCl2FN2 | |
1 g | |
4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro | |
DGMIGAHDDPJOPN-UHFFFAOYSA-N | |
4,6-dichloro-5-fluoropyrimidine | |
11286675 | |
98% |
213265-83-9 | |
91°C (196°F) | |
MFCD08056331 | |
UN3265 | |
Slightly soluble in water. | |
FC1=C(Cl)N=CN=C1Cl | |
166.96 | |
166.97 | |
Liquid |
Chemical Identifiers
213265-83-9 | |
166.96 | |
DGMIGAHDDPJOPN-UHFFFAOYSA-N | |
11286675 | |
FC1=C(Cl)N=CN=C1Cl |
C4HCl2FN2 | |
MFCD08056331 | |
4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro | |
4,6-dichloro-5-fluoropyrimidine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only