missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dichloro-5-fluoropyrimidine, 98%

Manufacturer:  Alfa Aesar™ H6177006

 View more versions of this product

Catalog No. AAH6177006



Description

Description

Applications
4,6-Dichloro-5-Fluoropyrimidine is an useful intermediate for pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

4,6-Dichloro-5-fluoropyrimidine
5g
C4HCl2FN2
UN3265
DGMIGAHDDPJOPN-UHFFFAOYSA-N
4,6-dichloro-5-fluoropyrimidine
11286675
Liquid
50°C to 60°C (14mmHg)
213265-83-9
MFCD08056331
4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro
C1=NC(=C(C(=N1)Cl)F)Cl
166.964
166.97
98%

Chemical Identifiers

213265-83-9
166.964
DGMIGAHDDPJOPN-UHFFFAOYSA-N
11286675
C1=NC(=C(C(=N1)Cl)F)Cl
C4HCl2FN2
MFCD08056331
4,6-dichloro-5-fluoro-pyrimidine, pyrimidine, 4,6-dichloro-5-fluoro, 4,6-dichloro-5-fluoro-1,3-diazine, pubchem18606, acmc-209fj8, ksc494o9j, 4,6-dichloro-5-fluoropyrimidine;, 4,6-dichloro-5-fluoro-pyrimidine;, 5-fluoro-4,6-dichloropyrimidine, 4 3h-pyrimidinone,2-chloro-5-fluoro
4,6-dichloro-5-fluoropyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Documents