missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine, 95%

Manufacturer:  Alfa Aesar™ H61166MD

 View more versions of this product

Catalog No. AAH61166MD



Specifications

Specifications

4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine
150728-13-5
MFCD03839838
IZGOBGVYADHVKH-UHFFFAOYSA-N
4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
10545706
Solid
250mg
C15H10Cl2N4O2
2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy
COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl
349.171
349.17
95%

Chemical Identifiers

150728-13-5
349.171
IZGOBGVYADHVKH-UHFFFAOYSA-N
10545706
COC1=CC=CC=C1OC2=C(N=C(N=C2Cl)C3=NC=CC=N3)Cl
C15H10Cl2N4O2
MFCD03839838
2,2'-bipyrimidine, 4,6-dichloro-5-2-methoxyphenoxy
4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents