missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dichloro-2-methylpyrimidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC434260050

 View more versions of this product

Catalog No. AC434260050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder or Crystals
≥96.0%
White to off-white
100.0
Glass bottle
97%
MFCD00090472
Authentic
163.01
5g
43° to 48°C
96.0
C5H4Cl2N2
1780-26-3
4, 6-Dichloro-2-methylpyrimidine

Chemical Identifiers

1780-26-3
100.0
C5H4Cl2N2
MFCD00090472
≥96.0%
96.0
97%
163.01
4, 6-Dichloro-2-methylpyrimidine

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning