missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,6-Dibromodibenzothiophene 98.0+%, TCI America™

$68.80 - $206.39

Chemical Identifiers

CAS 669773-34-6
Molecular Formula C12H6Br2S
Molecular Weight (g/mol) 342.048
MDL Number MFCD18451473
InChI Key ULGFJZPCGNTWFK-UHFFFAOYSA-N
Synonym 4,6-dibromo-dibenzothiophene, 4,6-dibromodibenzo b,d thiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo
PubChem CID 5245996
IUPAC Name 4,6-dibromodibenzothiophene
SMILES C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D4822200MG
View Documents
Tci America
D4822200MG
200mg
Each for $68.80
 
D48221G
View Documents
Tci America
D48221G
1g
Each for $206.39
 
Description

Description

Specifications

Chemical Identifiers

669773-34-6
342.048
ULGFJZPCGNTWFK-UHFFFAOYSA-N
5245996
C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
C12H6Br2S
MFCD18451473
4,6-dibromo-dibenzothiophene, 4,6-dibromodibenzo b,d thiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo
4,6-dibromodibenzothiophene

Specifications

White
200mg
C12H6Br2S
4,6-dibromo-dibenzothiophene, 4,6-dibromodibenzo b,d thiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo
C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
342.048
342.05
≥98.0% (GC)
186°C
669773-34-6
MFCD18451473
ULGFJZPCGNTWFK-UHFFFAOYSA-N
4,6-dibromodibenzothiophene
5245996
Crystal-Powder at 20°C
4,6-Dibromodibenzothiophene
Safety and Handling

Safety and Handling

TSCA : No

SDS