missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,6-Dibromodibenzothiophene, 96%

$245.20 - $749.47

Chemical Identifiers

CAS 669773-34-6
Molecular Formula C12H6Br2S
Molecular Weight (g/mol) 342.048
MDL Number MFCD18451473
InChI Key ULGFJZPCGNTWFK-UHFFFAOYSA-N
Synonym 4,6-dibromodibenzo b,d thiophene, 4,6-dibromo-dibenzothiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene
PubChem CID 5245996
IUPAC Name 4,6-dibromodibenzothiophene
SMILES C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66003MD View Documents Alfa Aesar™
H66003MD
250mg Each for $245.20
 
AAH6600303 View Documents Alfa Aesar™
H6600303
1g Each for $749.47
 
Specifications

Chemical Identifiers

669773-34-6
342.048
ULGFJZPCGNTWFK-UHFFFAOYSA-N
5245996
C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
C12H6Br2S
MFCD18451473
4,6-dibromodibenzo b,d thiophene, 4,6-dibromo-dibenzothiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene
4,6-dibromodibenzothiophene

Specifications

White
669773-34-6
MFCD18451473
ULGFJZPCGNTWFK-UHFFFAOYSA-N
4,6-dibromodibenzothiophene
5245996
Powder
4,6-Dibromodibenzothiophene
250mg
C12H6Br2S
4,6-dibromodibenzo b,d thiophene, 4,6-dibromo-dibenzothiophene, 6,10-dibromo-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, dibenzothiophene, 4,6-dibromo, 6,10-dibromo-8-thiatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene
C1=CC2=C(C(=C1)Br)SC3=C2C=CC=C3Br
342.048
342.05
96%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning